Would you like to log in again?
SP Bel-Art Dry-Keeper™ Horizontal Auto-Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
Dry-Keeper™
H42058-0003
74950-342EA 3254.56 USD InStock
74950-342
SP Bel-Art Dry-Keeper™ Horizontal Auto-Desiccator Cabinet, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
The 120VAC (11 watt) electronic dehumidification system maintains relative humidity levels at 30 to 40% (cabinet interior rH% levels can differ depending upon room temperature, room rH% and usage conditions) keeping dry air circulating throughout the cabinet.
  • Durable
  • Clear PVC panels
  • Sleek black ABS accents
  • Horizontal profile
  • Movable shelves for adjustable dry storage configurations
Permanent desiccant is contained in the control unit and a red light indicates operation of the 4½ hour absorption period while a green light indicates operation of the ½ hour regeneration of the desiccant.
The cabinet features a spacious 2 cu. ft. interior volume and a large, in-door dial hygrometer for easy visualization of rH%. Two adjustable, removable, and perforated shelves facilitate full circulation of air and shelf rails can be repositioned on any one of 8 steps set at 28 mm intervals. The gasketed door opens down and closes with two latches for a tight seal. Interior opening measures 45 x 23 cm (17¾ x 9").
Certifications: CE Marked.
Ordering information: Additional shelves and rails sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty