Would you like to log in again?
PolarSafe™ Stainless Steel Freezer Bin Storage Racks, Argos Technologies
Supplier: Argos Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PolarSafe®
04397-80 04398-00 04397-88 04397-90 04397-64 04397-92 04397-72 04397-76 04397-98 04398-01 04397-93 04397-91 04397-73 04397-69 4397-82 04398-03 04397-77 04397-63 04397-95 04397-65 04397-89 04397-84 04397-78 04397-71 4397-86 04397-81 04398-02 04397-66 04397-99 04397-96 04397-74 04397-97 04397-70 04397-68 04397-62 04397-87 04397-79 04397-94 04397-83 04397-67 4397-75 04397-85
75778-018EA 178.88 USD InStock
75778-018 75778-212 75778-036 75778-014 75778-058 75778-214 75778-016 75778-038 75778-022 75778-044 75778-186 75778-024 75778-200 75778-046 75778-188 75778-040 75778-062 75778-042 75778-020 75778-184 75778-060 75778-206 75778-208 75778-048 75778-026 75778-202 75778-204 75778-028 75778-198 75778-032 75778-054 75778-210 75778-034 75778-056 75778-012 75778-194 75778-050 75778-196 75778-052 75778-030 75778-190 75778-192
PolarSafe™ Stainless Steel Freezer Bin Storage Racks, Argos Technologies
Boxes Cryoboxes
PolarSafe™ Stainless Steel Bin Storage Racks have been designed to store miscellaneous bottles, tubes, vials, bags, or boxes that will not fit in standard storage racks
Stainless steel construction with integrated dual handles. Supplied with a lifetime warranty.
Both rack sides are made from wire mesh to allow for easy viewing of contents. The bin storage racks are available in fixed or drawer rack styles. The drawer racks are fully framed and work just like any standard single drawer rack. The open format works well with –20⁰C freezers, or larger walk-in freezers with adjustable shelving.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty