PolarSafe™ Stainless Steel Bin Storage Racks have been designed to store miscellaneous bottles, tubes, vials, bags, or boxes that will not fit in standard storage racks
Stainless steel construction with integrated dual handles. Supplied with a lifetime warranty.
Both rack sides are made from wire mesh to allow for easy viewing of contents. The bin storage racks are available in fixed or drawer rack styles. The drawer racks are fully framed and work just like any standard single drawer rack. The open format works well with –20⁰C freezers, or larger walk-in freezers with adjustable shelving.