Would you like to log in again?
Top Taper Inlet Liner with Wool for Bruker/Varian GCs Equipped with 1078/1079 Inlets, Restek
Supplier: Restek
20901-213.5 20901-211
75778-270PK 280.83 USD InStock
75778-270 75778-324
Top Taper Inlet Liner with Wool for Bruker/Varian GCs Equipped with 1078/1079 Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty