Would you like to log in again?
Dual Column mini-Lam Direct Injection Tee Kit for ¹/₄" packed column inlets, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20436
75778-664EA 242.71 USD InStock
75778-664
Dual Column mini-Lam Direct Injection Tee Kit for ¹/₄" packed column inlets, Restek
Fittings and Connectors
Inverted cup design offers complete sample vaporization.
  • Permits larger sample volumes
  • Incorporates a Press-Tight® taper in each outlet leg, to make a perfect, dead-volume-free connection to each analytical column (ODs ranging from 0.4 to 0.8 mm)
  • Allows visual confirmation of the column connection
Open-top design makes it easy to pack with glass wool, keeping sample residue from contaminating the cup.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty