Would you like to log in again?
Base-Deactivated Glass Wool, Restek
Supplier: Restek
20999
75778-742EA 90.81 USD InStock
75778-742
Base-Deactivated Glass Wool, Restek
Glass and Cotton Wool
Ideal for amines and other basic compounds.
Base-deactivated glass wool produces high and reproducible responses for basic compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty