Would you like to log in again?
INLET LINER STRAIGHT 4X78.5X6.5MM GC PK5
Supplier: Avantor
Straight Inlet Liner Splitless Liner^ Straight Inlet Liner, Splitless Liner, Injector Liner for GCs, Liner Inside (ID) diameter: 4.0 mm, Length: 78.5 mm, Outside (OD) diameter: 6.5 mm, Deactivation: Base Deactivated, Packing: Deactivated Wool, Material: Borosilicate Glass
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INLET LINER STRAIGHT 4X78.5X6.5MM GC PK5 75778-866   Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty