Would you like to log in again?
Drilled Uniliner® Inlet Liner with Hole near Top for Shimadzu 17A, 2010 and 2014 GCs equipped with split/splitless inlets, Restek
Supplier: Restek
Uniliner®
21286 21286-214.5
75779-322PK 417.04 USD InStock
75779-322 75779-116
Drilled Uniliner® Inlet Liner with Hole near Top for Shimadzu 17A, 2010 and 2014 GCs equipped with split/splitless inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty