Would you like to log in again?
PROSAT® Sterile Low Endotoxin Knitted Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PROSAT®
PSWE0002 PSWE0003 PSWE0001
75779-606CS 1263.05 USD InStock
75779-606 52428-882 75833-990
PROSAT® Sterile Low Endotoxin Knitted Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
PROSAT® Sterile Polynit Heatseal LE Wipes are made of 100% knitted polyester Polynit Heatseal, presaturated with a blend of 70% isopropanol and 30% water for injection (WFI). Each lot is tested before release and low endotoxin certified to less than 1 EU/wipe.
  • Recommended for Grade A/B or ISO Class 5 environments (product contact areas)
  • 100% knitted polyester fabric results in very low levels of particles and fibers
  • Laser sealed edges reduce fiber generation from the edge of the wipe
  • Guaranteed endotoxin levels of 1EU/device make them ideal for product contact surfaces
  • Small number of wipes per resealable pouch to eliminate waste
  • Presaturated wipe reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels
The sealed edge wipe produces very low levels of particles and fibers and is ideal for use on product contact surfaces. Each resealable pouch contains a small number of wipes, eliminating any waste at the end of a session. Each pouch is individually bagged for easy entry into the sterile suite. Small footprint, hanging pouches available, ideal for use in isolator and RAB systems.
Presaturated wipes provide many benefits including solvent control, reduced Volatile Organic Compounds (VOCs), and increased process control and repeatability. The resealable pouch reduces waste. PROSAT® Sterile Polynit Heatseal LE Wipes are validated sterile and suitable for use in aseptic suites and the most critical pharmaceutical cleanrooms.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty