Would you like to log in again?
PLATE ADAPTER M30 THREAD 15 L HEADPLATE
Supplier: Chemglass
Adapter Plate M30^ Adapter Plate, M30, for 15L Bioreactor to M30, Allows for the conversion of the 15 liter headplate from a 69mm opening to an M30 thread, thereby accepting the CLS-1385-35 agitator assembly
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE ADAPTER M30 THREAD 15 L HEADPLATE 75780-170 CLS-1385-30 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty