Would you like to log in again?
Bioreactor Complete Accessories Kit
Supplier: Chemglass
CLS-1390-01
75780-186EA 437.97 USD InStock
75780-186
Bioreactor Complete Accessories Kit
Chromatography Supplies Chromatography Inlet Liners
This pre fabricated, single use bio reactor kit consists of the 8 components. They are packed in an ISO Class 7 clean room, double bagged and ready for use.
  • Minimize labor costs
  • Eliminates cleaning validation
  • Reduces set up time and costs
  • Minimize injuries associated with cutting in-house
  • Improved consistency
  • Gives the ability to assemble in open lab type environment
  • Provides aseptic solution to close off an unused line
  • Autoclave maxium 15 psi @ 134 °C, 60 min
  • Gamma radiation 25 kGy to 50 kGy
Kits are fabricated with full traceability and accompanying validation documentation, which meets USP Class VI requirements. These pre-assembled kits can be customized to any validated components currently specified on your bioreactor design.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty