Would you like to log in again?
Miriad Rapid HCV/HIV Antibody Test POU+, MedMira
Supplier: MedMira
Miriad™
815311006012
75780-214BX 657.18 USD InStock
75780-214
Miriad Rapid HCV/HIV Antibody Test POU+, MedMira
Clinical Diagnostic Supplies Clinical Reagents and Kits
Miriad Rapid HCV/HIV Antibody Test employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of Hepatitis C and HIV-1/2
Miriad HCV/HIV features a built-in reagent control line, works with whole blood, serum, and plasma specimens, and requires no additional equipment. Test kits can be stored for 12 months at 2–30°C (35.6–86°F).
In three easy steps users can obtain multiple instant results on a single test cartridge.
Ordering information: Each box of Miriad HCV/HIV POU+ Format includes 25 unitized test kits containing 1 test cartridge, 1 InstantGold cap, 1 Buffer Vial 1, 1 Buffer Vial 2, 1 transfer pipette and a package insert.
HBc/HIV/HCV Antibody Test controls are available as an accessory to this test..
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty