Would you like to log in again?
Biox Cardboard Freezer Boxes
Supplier: TradeWinds (TWD)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Biox
BX90-2381 BX90-2300 BX90-2200 BX90-2281
75780-330CS 383.8 USD InStock
75780-330 75780-332 75780-312 75780-314
Biox Cardboard Freezer Boxes
Boxes Cooling/Freezing Boxes and Bags
Premium cardboard freezer boxes with a water-resistant coating.
  • Chemically resistant to alcohols and mild organic solvents
  • Temperature range: –196 to 121°C (–320 to 250 °F)
  • Accommodates 0.5 ml, 1.5 ml, or 2.0 ml microtubes
  • Suitable for liquid nitrogen storage
Boxes feature a water-resistant coating for improved durability.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty