Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgY Goat Polyclonal Antibody (Horseradish Peroxidase (HRP))

Supplier: Immunoreagents
GTXTK-003-DHRPX
75783-564EA 156.2 USD InStock
75783-564
Anti-IgY Goat Polyclonal Antibody (Horseradish Peroxidase (HRP))
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Horseradish Peroxidase (HRP)
Epitope:
Host: Goat
Isotype: IgY
Reactivity: Turkey

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty