Would you like to log in again?
VWR® Traceable® Excursion-Trac™ USB Datalogging ULT Freezer Thermometers
Supplier: Avantor
VWR®
90013-65 93679-82
75784-088EA 428.58 USD InStock
75784-088 75784-090
VWR® Traceable® Excursion-Trac™ USB Datalogging ULT Freezer Thermometers
Data Loggers
USB Datalogging ULT freezer thermometer with user-defined time intervals.
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Smart-Alarm™ features a visual/audible alarm that continues to alarm even if the unit returns to non-alarm conditions
  • Stores up to ten unique alarm events
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data - unit can remain in use while downloading and analyzing data
  • Status indicators - Low battery, memory full, USB data transfer and active alarm state
  • Smart-Alarm™ signals out-of-range conditions
Hi/Lo Alarms and time/date stamps: Data for up to ten different alarm events can be retrieved with the most current event viewable on the display. With exportable data, A/C plug and temperature minimum/maximum memory.
Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range. Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments. Data for up to 10 different alarm events can be retrieved with the most current event viewable on the display.
When an out-of-range condition is sensed, the Smart-Alarm™ triggers. Smart-Alarm™ features visual and audible indicators which signal continuously until acknowledged, even if the parameter returns to the non-alarm range. Meets current CDC requirements for vaccine storage and monitoring. Thermometer has cable length of 10' with calibration points of −80, −40, 0, 100 °C. LCD digit size is ¹/₂".
New feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac using a USB flash drive (not included). No software is needed for computer interface. Each thermometer has a unique ID allowing multiple units to be used in the same location. Datalogging features has monitor readings overnight, on weekends, or for any time period with rolling data log. User-defined datalogging interval of one observation/minute to one observation/24 hours. Memory capacity maintains a year of recorded data when using one minute intervals and longer when using larger intervals. Highly accurate and reliable. Ideal for monitoring temperatures in refrigerators, freezers, water baths, heating blocks, and incubators.
Certifications: Traceable® Certificate. Multi-point calibration on an individually-numbered Traceable® Certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA.  It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. CE marked.
Ordering information: Supplied with platinum RTD sensors and stainless steel 316 probes, stand, Velcro™, magnetic strips, wall mount, batteries and Traceable® Certificate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty