Would you like to log in again?
PLATE DEEP 96 WELL SQ AUTCLV PP CLR 2ML
Supplier: Corning
Deep Well Plate 2ml^ Deep Well Plate, Pre-Sterilized, Square 96 Well Individually Wrapped, Size: 2.0ml, Color: Clear, Material: Polypropylene, High Profile, Single Wrapped, Storage Microplate, Guaranteed RNase, DNase and pyrogen free, Autoclavable
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE DEEP 96 WELL SQ AUTCLV PP CLR 2ML 75784-550 P-2ML-SQ-BC-RUCDR Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty