Would you like to log in again?
OC-Light® S FIT Test Kits, Polymedco
Supplier: Polymedco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
OC-Light®
FBPU-BULKS FBPU-ABRMP120S FBPU-ADRS FOBBTLS FBPUS FBPU-NBRMPS FBPUCUSTOM120S FOBSTRS FOB50S
75784-750KT 882.55 USD InStock
75784-750 75784-762 75784-760 75784-754 75784-578 75784-752 75784-764 75784-758 75784-756
OC-Light® S FIT Test Kits, Polymedco
Clinical Diagnostic Supplies Clinical Reagents and Kits
OC-Light S FIT is a manual fecal immunochemical test for the detection of blood in stool that can lead to colorectal cancer.
  • Simple strip procedure with qualitative results in 5 minutes
  • Utilizes colloidal gold to enhance test line intensity and clarity
  • Requires no dietary or medicinal restrictions
Enables the physician or the patient to easily collect a sample and return the sample to the lab or physician office for quick and accurate results. Increased sensitivity, specificity, and patient compliance equates to less false positive results and more of the right patients being referred to colonoscopy.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty