Would you like to log in again?
ITSE Animal Free®, InVitria
Supplier: InVitria
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
777ITS03210X10 777ITS032-10ML 777ITS032100ML
75784-766EA 1322.03 USD InStock
75784-766 10847-794 10847-714
ITSE Animal Free®, InVitria
Cell Culture Supplements and Additives
InVitria’s ITS, ITSE, and ITSE+A Animal-Free® are recombinant, blood-free cell culture media supplements used to optimize cell growth.
For optimal proliferation and performance, cells cultured must be supplied with a critical proteins and components:
Insulin: promotes uptake of glucose and amino acids, facilitating cellular signaling
Transferrin: delivers intracellular iron via the transferrin receptor, drives cellular respiration, chelates free iron, and reduces generation of cytotoxic free radicals and peroxide
Selenium: required for the activity of antioxidant enzymes such as glutathione peroxidase and thioredoxin reductase
Ethanolamine: phospholipid precursor that improves the performance of cells in serum-free media and is required for the growth of certain cell types
Albumin: binds, transports, and delivers ions, fatty acids, vitamins, hormones, small-molecule drugs, and toxins; regulates serum pH and osmotic pressure; and provides antioxidant activity
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty