Would you like to log in again?
Essentials Spot Indole Reagent, Pro-Lab Diagnostics
Supplier: Pro-Lab Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PL.850-10 PL.850-20 PL.850-30 PL.391-10 PL.375 PL.850-3 PL.701
75784-898EA 115.2 USD InStock
75784-898 75784-912 75784-910 75784-900 75784-916 75784-906 75784-914
Essentials Spot Indole Reagent, Pro-Lab Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Pro-Lab’s Spot Indole Reagent is to be used in the qualitative method to determine the ability of an organism to split indole from the tryptophan molecule.
  • Pro-Lab Mucolyse PL.701 enhances the isolation of organisms responsible for chronic lung disease
  • PYR test for the determination of Pyrrolidonyl peptidase (PYRase) activity
  • PYR cards contain a substrate used for detection of PYRase activity
  • Rabbit Coagulase Plasma for the qualitative detection of the coagulase enzyme produced by Staphylococcus aureus
The amino acid tryptophan can be oxidized by certain bacteria using intracellular enzymes collectively called ‘tryptophanase’, resulting in the production of indole
Pro-Lab Mucolyse PL.701 is a sputum liquefying agent used to digest and thin out the sputum, thus enhancing the isolation of organisms responsible for chronic lung disease.
The Prospot™ PYR test is a rapid colourimetric test for the determination of Pyrrolidonyl peptidase (PYRase) activity.
Prospot™ PYR test cards contain a substrate used for detection of PYRase activity. When the colour developer is added, the enzymatic hydrolysis of the substrate by enterococci or Group A streptococci produces a red colour.
The indole is detected by the p-dimethylaminocinnamaldehyde, which involves a chemical combination producing a distinct blue colour. The presence or absence of indole formation is used for bacterial identification.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty