Would you like to log in again?
Microbank Cryoblock, Pro-Lab Diagnostics
Supplier: Pro-Lab Diagnostics
PL.156 PL.155-1
75784-934PK 105.07 USD InStock
75784-934 75784-932
Microbank Cryoblock, Pro-Lab Diagnostics
Block Heaters Blocks
A 20 well, insulated cryoblock is available which enables the operator to remove a number of Microbank™ vials from the freezer for sub culture without risk of the culture beads defrosting.
The cryoblock is insulated with a polystyrene shield.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty