Would you like to log in again?
AmnioTest™, Pro-Lab Diagnostics
Supplier: Pro-Lab Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AmnioTest™
PL.901-20 PL.902 PL.911 PL.910 PL.913 PL.915 PL.914 PL.912 PL.901
75784-960EA 98.52 USD InStock
75784-960 75784-952 75784-950 75784-956 75784-946 75784-954 75784-944 75784-958 75784-948
AmnioTest™, Pro-Lab Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
AmnioTest™ is a qualitative, pH-based, swab screening system intended as an aid in detecting rupture of amniotic membrane in pregnant women.
  • Detect rupture of amniotic membrane in pregnant women
  • Determine the presence of amniotic fluid
  • Swab absorbs fluid associated with the tissue and the dye develops a colour which correlates with the pH of the absorbed fluid
Rupture of the amniotic membrane can result in small volumes of amniotic fluid leaking into the upper vagina
A swab impregnated with nitrazine yellow dye is brought into contact with the upper vagina. The swab absorbs fluid associated with the tissue and the dye develops a colour which correlates with the pH of the absorbed fluid over the range pH 5.0 to pH 7.5. Amniotic fluid has a neutral pH while the pH of the upper vagina is normally acidic. A pH of 6.5 or higher in the upper vagina is consistent with leakage of amniotic fluid.
The presence of amniotic fluid tends to elevate the pH of the upper vagina. Detection of this pH increase using a pH indicator dye has been shown to assist in determining the presence of amniotic fluid.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty