Would you like to log in again?
McFarland Standard, Pro-Lab Diagnostic
Supplier: Pro-Lab Diagnostics
SD2304 SD2300 SD2350 SD2302 SD2301 SD2303
75785-176EA 233.9 USD InStock
75785-176 75785-178 75784-924 75785-180 75784-928 75784-926
McFarland Standard, Pro-Lab Diagnostic
Media for Microbiology Dehydrated Media for Microbiology
The McFarland Standards are intended for adjusting densities of bacterial suspensions and have a shelf life of up to two years from the date of manufacture.
Delivery information: All standards are supplied in a protective carrying case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty