Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Salmonella O Group Antisera, Pro-Lab Diagnostics
Supplier: Pro-Lab Diagnostics
PLA044 PLA048 PLA041 PLA032 PLA051 PLA028 PLA046 PLA057 PLA043 PLA038 PLA036 PLA062 PLA054 PLA039 PLA034 PLA058 PLA061 PLA059 PLA045 PLA060 PLA035 PLA029 PLA021 PLA026 PLA024 PLA049 PLA023 PLA033 PLA047 PLA053 PLA025 PLA027 PLA042 PLA040 PLA052 PLA056 PLA031 PLA022 PLA030 PLA037 PLA055
75785-252EA 361.99 USD InStock
75785-252 75785-296 75785-274 75785-230 75785-276 75785-232 75785-298 75785-254 75785-234 75785-256 75785-278 75785-258 75785-236 75785-290 75785-292 75785-270 75785-272 75785-294 75785-250 75785-228 75785-264 75785-242 75785-286 75785-288 75785-244 75785-266 75785-268 75785-246 75785-224 75785-248 75785-226 75784-978 75785-280 75785-282 75785-260 75785-240 75785-262 75785-284 75784-982 75784-980 75785-238
Salmonella O Group Antisera, Pro-Lab Diagnostics
Microbiology Test Systems Antibiotic Tests
Salmonella O antisera is specific for slide agglutination testing.
  • Shelf lives up to five years from date of manufacture
  • Supplied in color coded dropper bottles
The range includes a full panel of polyvalent somatic antisera, monovalent somatic antisera, polyvalent flagella antisera and monovalent flagella antisera for traditional slide agglutination testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty