Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Thermal Mixers, Thermo Scientific

Supplier: Thermo Scientific
13687721 13687718 13687711 13687717 13687720 13687719
75786-282EA 4582.82 USD InStock
75786-282 75786-280 75786-268 75786-288 75786-286 75786-284
Thermal Mixers, Thermo Scientific
Shakers and Mixers Shaking Incubators
The Thermo Scientific Thermal Mixer offers variable speed and temperature control with a choice of blocks for microtubes and microplates.

  • Fast heat-up and cooling times and quick to reach specified mixing speed
  • Temperature range 4°C to 100°C
  • Temperature control/set range 15°C below ambient to 100°C
  • Shaking speed 250 to 1400 rpm
  • Temperature stability ±0.1°C
  • Even mixing across the thermoshaker block
  • Excellent uniformity across the block
  • Digital timer with sound alarm 1 min to 96 hours
  • Temperature calibration function – Compensate for differences in the thermal behavior of different tubes and samples
  • Choice of 5 interchangeable blocks
  • LCD Display indicates the set and actual time of operation, mixing speed, and temperature
  • Heating/Cooling and mixing functions can be run independently of each other

Heating and cooling capability to cover a wide range of applications. Compact size with quiet operation. Convenient packages include Thermal Mixer with a block.

Certifications: CE compliant.

Ordering information: 2 year warranty.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty