Would you like to log in again?
LBP, Soluble ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-304-KI01
75786-320EA 1230 USD InStock
75786-320
LBP, Soluble ELISA Kit, Enzo Life Sciences
Assays ELISAs
Quantitative determination of human, dog, pig, rabbit, bovine and horse-soluble LBP levels.
For the quantitative determination of human, dog, pig, rabbit, bovine and horse soluble LBP levels in serum, plasma and cell culture medium.
Ordering information: Contains 1 precoated ELISA module1 vial anti-LBP (human) detecting MAb1 vial LBP standard (human)1 vial reference serum2 tablets PBS1 vial dilution buffer1 vial Tween 201 vial stopping solution1 vial substrate solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty