Would you like to log in again?
FSH ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIT108-0001
75786-330EA 294 USD InStock
75786-330
FSH ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive FSH ELISA kit enabling detection of FSH in serum, plasma and tissue culture media in just 2 hours.
  • Sensitive measurement of FSH, detecting as little as 0.5 mIU/ml
  • Negligible reactivity from similar glycoprotein hormones
  • High throughput format with results in two hours for up to 37 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of FSH in tissue culture media, serum and plasma in human samples.
The FSH ELISA kit is a colorimetric sandwich immunoassay kit with results in two hours.
Ordering information: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, Substrate, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty