Would you like to log in again?
ACTH ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT138-0001
75786-336EA 702 USD InStock
75786-336
ACTH ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ACTH ELISA kit enabling detection of ACTH in human plasma in 4.5 hours.
  • Sensitive measurement of ACTH, detecting as little as 0.46 pg/ml
  • Negligible reactivity to similar hormones
  • High throughput with results in 4.5 hours
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ACTH in plasma in human samples.
The ACTH ELISA kit is a colorimetric immunoassay kit with results in 4.5 hours.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty