Would you like to log in again?
IL-5 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT139-0001
75786-338EA 891.14 USD InStock
75786-338
IL-5 ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive IL-5 ELISA kit enabling detection of IL-5 in serum, plasma, urine and cell culture supernates in just 3 hours.
  • Sensitive measurement of IL-5, detecting as little as 2 pg/ml
  • Negligible reactivity to related proteins
  • High throughput with results in three hours
  • Fully quantitative results that pass semi-quantitative Western blot analysis
For the quantitative determination of IL-5 in cell culture supernates, serum, plasma (Heparin & EDTA), urine, in human samples.
The IL-5 ELISA kit is a colorimetric sandwich immunoassay kit with results in three hours.
Ordering information: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Substrate, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty