Would you like to log in again?
Methotrexate ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT142-0001
75786-344EA 708 USD InStock
75786-344
Methotrexate ELISA Kit, Enzo Life Sciences
Assays ELISAs
Ultra sensitive Methotrexate ELISA kit enabling detection of Methotrexate in serum, plasma, and urine in just 1.5 hours.
  • Sensitive measurement of methotrexate, detecting as little as 0.087 ng/ml
  • Broad dynamic range suitable for a large variety of samples
  • High throughput format with results in 1.5 hours for up to 38 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of methotrexate in serum, plasma and urine in human, mouse, and rat samples.
The Methotrexate ELISA kit is a colorimetric competitive immunoassay kit with results in 1.5 hours.
Ordering information: Contains Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty