Would you like to log in again?
Comply™ Chemical Integrators, 3M™
Supplier: 3M
MMM41360 MMM1243B MMM1243RE
75787-962CS 293.45 USD InStock
75787-962 75787-964 75786-420
Comply™ Chemical Integrators, 3M™
Sterilization Indicators
Comply™ Chemical Integrators provides an immediate, accurate, and easy-to-read method of monitoring sterilization process conditions inside each pack.
It can be used in all steam sterilizers. The easy-to-read moving front style has been shown to have a higher reading accuracy than color interpretation style indicators.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty