Would you like to log in again?
TGF-alpha ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT132-0001
75786-644EA 792 USD InStock
75786-644
TGF-alpha ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ELISA kit enabling detection of TGF-α.
  • Sensitive measurement of TGF-α, detecting as little as 9.375 pg/ml
  • Negligible reactivity from similar growth factors
  • High throughput format with results in 3.5 hours
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of TGF-α in serum, plasma, cell culture supernatant, tissue homogenates in human samples.
The TGF-α ELISA kit is a colorimetric immunometric immunoassay kit.
Ordering information: Contains Microtiter Plate, Standard, Detection Antibody, Detection Antibody Diluent, Sample Diluent, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty