Would you like to log in again?
Peptide YY (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT133-0001
75786-646EA 868 USD InStock
75786-646
Peptide YY (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive Peptide YY ELISA kit enabling detection of PYY in serum.
  • Sensitive measurement of PYY, detecting as little as 18.75 pg/ml
  • Negligible reactivity from similar peptides
  • High throughput format with results in 3.5 hrs
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of Peptide YY in serum, plasma, cell culture supernatant and tissue homogenate in human samples.
The Peptide YY ELISA kit is a colorimetric immunometric immunoassay kit with results in 3.5 hours.
Ordering information: Contains Microtiter Plate, Standard, Sample Diluent, Detection Antibody, Wash Buffer Concentrate, Conjugate, Conjugate Diluent, Substrate, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty