Would you like to log in again?
ApoE ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT134-0001
75786-648EA 851.53 USD InStock
75786-648
ApoE ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ApoE ELISA kit enabling detection of ApoE in serum, plasma, cell culture supernates, and tissue homogenates in 3.5 hours.
  • Sensitive measurement of ApoE, detecting as little as 14.063 pg/ml
  • Negligible reactivity to similar proteins
  • High throughput with results in 3.5 hours
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ApoE in serum, plasma, cell culture supernates and tissue homogenates in human samples.
The ApoE ELISA kit is a colorimetric immunometric immunoassay kit with results in 3.5 hrs.
Ordering information: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Conjugate Buffer Substrate, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty