Would you like to log in again?
Tegaderm™ High Performance Foam Non-Adhesive Dressing, 3M™
Supplier: 3M
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MMM90600 MMM90605 MMM90604 MMM90603 MMM90602
75786-804CS 1423.33 USD InStock
75786-804 75786-802 75786-800 75786-798 75786-796
Tegaderm™ High Performance Foam Non-Adhesive Dressing, 3M™
First Aid Supplies Wound Dressings
Non-adhesive foam dressing that effectively handles low to high exudating wounds.
  • High absorbency and high breathability reduce the risk of maceration
  • Demonstrates high fluid handling capacity under compression
  • Dressing does not stick to wound bed, minimizing disruption of healing tissue
Innovative layer technology absorbs and evaporates moisture to maintain an optimal wound healing environment.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty