Would you like to log in again?
Arizant Bair Paws™ Flex Warming Gowns, 3M™
Supplier: 3M
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MMM84003 MMM83203 MMM81103 MMM83003 MMM84103 MMM84203 MMM83103 MMM81203 MMM81003
75786-936CS 1671.8 USD InStock
75786-936 75786-926 75786-934 75786-932 75786-930 75786-942 75786-940 75786-938 75786-928
Arizant Bair Paws™ Flex Warming Gowns, 3M™
Disposable Garments
Arizant Bair Paws™ Flex Gown combines the comfort and convenience of the Bair Paws™ gown with Bair Hugger™ forced-air warming blankets.
  • Improve patient satisfaction by keeping patients warm, cozy and covered
  • Warming tools built right into the gown including a head drape, tie strips, and a patient adhesive strip
  • Small size gown is perfect for warming smaller surgical patients throughout the perioperative process
  • Accommodates adolescent and petite surgical patients
With a single gown, clinicians can warm during virtually any procedure, including those involving upper, torso, or lower body warming. Two inserts allow for greater warming flexibility in clinical or comfort warming procedures. Upper warming insert includes all the features of a Bair Hugger™ upper body blanket. Separate upper and lower inserts allow users to activate warming only where needed.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty