Would you like to log in again?
Ioban™ 2 Antimicrobial Incise Drape, 3M™
Supplier: 3M
Ioban™ 2
MMM6651EZ MMM6661EZ MMM6648EZ
75787-724CS 1728.56 USD InStock
75787-724 75787-726 75787-722
Ioban™ 2 Antimicrobial Incise Drape, 3M™
First Aid Supplies Surgical Drapes
Unique framed delivery system enables simplified, one-person application.
Size is ideally suited for high-volume, minimally invasive implant procedures. Iodophor-impregnated adhesive provides continuous, broad-spectrum antimicrobial activity, and superior adhesion to the skin and wound edge. Strong, conformable, and breathable polyester incise film provides a sterile surface, and helps to decrease bacterial contamination from skin to gloves, gowns, instruments, and implants.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty