Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Precise™ Multi-Shot MS Disposable Skin Stapler, 3M™
Supplier: 3M
MMMDS-15
75788-008CS 903.89 USD InStock
75788-008
Precise™ Multi-Shot MS Disposable Skin Stapler, 3M™
Staplers Staple Guns
Precise™ Multi-Shot MS disposable skin stapler dispenses box-style staples.
  • Quick and easy to use
  • Designed to be a versatile, reliable, and cost-effective wound closure tool
These "floating" staples maintain wound edge apposition without the need to fill them with tissue, allowing for appropriate edema to occur.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty