Would you like to log in again?
Curos Tips™ Disinfecting Cap Strip for Male Luers, 3M™
Supplier: 3M
MMMCM5-200
75788-304CS 1861.24 USD InStock
75788-304
Curos Tips™ Disinfecting Cap Strip for Male Luers, 3M™
Closures Safety Caps and Safety Waste Caps
Curos Tips™ use 70% isopropyl alcohol (IPA) to passively disinfect and protect all critical surfaces of male luer connections on intravenous tubing sets.
  • Helps reduce the risk of contaminants from entering the catheter post-insertion
  • Fits majority of male luer connections
  • Disinfects critical surfaces of male luer connections in 1 minute
  • Packaged to hang from I.V. pole for easy access, greater compliance, and reduced waste
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty