Synonyms:
Dihydrogen oxide
CCPW-0125
CCPW-2000
CCPW-0500
CCPW-020Q
CCPW-010L
CCPW-1000
75788-538CS
693.87
USD
InStock
75788-538
10218-876
75788-536
10218-880
10220-414
75788-540
Water, Cell Culture Grade
Water
Distilled cell culture grade water.
- Prepared by distillation
- Steam sterilization in final packaging
- 0.1 micron sterile filtration
Distilled cell culture grade water, <0.005EU/mL. Prepared by distillation, 0.1 micron sterile filtration and steam sterilization in final packaging.