Would you like to log in again?
Human Recombinant NAD Kinase (from E. coli)
Supplier: ProSci
91-250
75788-598EA 946.47 USD InStock
75788-598
Human Recombinant NAD Kinase (from E. coli)
Enzymes
NAD Kinase (NADK) is an enzyme that belongs to the NAD Kinase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
It is a widely expressed enzyme, but it is not detected in skeletal muscle. NADK converts Nicotinamide Adenine Dinucleotide (NAD+) into NADP+, through phosphorylating the NAD+ coenzyme. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis. The structure of the NADK from the archaean Archaeoglobus fulgidus has been determined.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty