Would you like to log in again?
Human Recombinant Ketohexokinase (from Cells)
Supplier: ProSci
91-843
75788-632EA 744.04 USD InStock
75788-632
Human Recombinant Ketohexokinase (from Cells)
Enzymes
Ketohexokinase, also known as Hepatic fructokinase, is a member of the carbohydrate kinase PfkB family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It exits as a homodimer and most abundant in liver, kidney, gut, spleen and pancreas. Low levels also found in adrenal, muscle, brain and eye.This enzyme catalyzes conversion of fructose to fructose-1-phosphate. It is the first enzyme with a specialized pathway that catabolizes dietary fructose. Defects in KHK are the cause of fructosuria.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty