Would you like to log in again?
Human Recombinant Inosine Triphosphatase (from E. coli)
Supplier: ProSci
91-989
75788-652EA 946.47 USD InStock
75788-652
Human Recombinant Inosine Triphosphatase (from E. coli)
Enzymes
Inosine Triphosphate Pyrophosphatase (ITPase) is a cytoplasmic enzyme that belongs to the HAM1 NTPase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
ITPase hydrolyzes the non-canonical purine nucleotides inosine triphosphate (ITP) and deoxyinosine triphosphate (dITP) to the monophosphate nucleotide (IMP) and diphosphate. The ITPase enzyme acts as a homodimer and does not distinguish between the deoxy- and ribose forms. ITPase probably excludes non-canonical purines from RNA and DNA precursor pools, thus preventing their incorporation into RNA and DNA and avoiding chromosomal lesions. Defects in ITPase is thought to be inherited and is characterized by an over-accumulation of ITP in erythocytes, leukocytes and fibroblasts.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty