Would you like to log in again?
Human Recombinant Glycine N-Methyltransferase (from E. coli)
Supplier: ProSci
91-999
75788-672EA 946.47 USD InStock
75788-672
Human Recombinant Glycine N-Methyltransferase (from E. coli)
Enzymes
Glycine N-Methyltransferase (GNMT) is a tetrameric cytosolic protein
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
GNMT catalyzes the synthesis of N-methylglycine from glycine using S-adenosylmethionine (AdoMet) as the methyl donor. It can affects DNA methylation by regulating the ratio of S-adenosylmethionine to S-adenosylhomocystine, playing an important role in maintaining normal AdoMet levels. GNMT is highly expressed in liver. As a major folate-binding protein, GNMT takes part in the detoxification pathway. Defects in GNMT are the cause of hypermethioninemia. the patients with this deficiency are mild hepatomegaly and chronic elevation of serum transaminases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty