Would you like to log in again?
Human Recombinant Methylmalonyl-CoA epimerase (from Cells)
Supplier: ProSci
92-387
75788-702EA 464.46 USD InStock
75788-702
Human Recombinant Methylmalonyl-CoA epimerase (from Cells)
Enzymes
Methylmalonyl-CoA epimerase, mitochondrial(MCEE)is an enzyme which belongs to the glyoxalase I family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It converts (S)-methylmalonyl-CoA to the (R) form, catalyses the following chemical reaction: (R)-methylmalonyl-CoA (S)-methylmalonyl-CoA. It plays an important role in the catabolism of fatty acids with odd-length carbon chains. This protein deficiency is an autosomal recessive inborn error of AA metabolism, involving valine, threonine, isoleucine and methionine. This organic aciduria can appear in the neonatal period with life-threatening metabolic acidosis, hyperammonemia, feeding difficulties, pancytopenia and coma.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty