Would you like to log in again?
Human Recombinant Carbonic Anhydrase 1 (from E. coli)
Supplier: ProSci
91-090
75788-904EA 533.61 USD InStock
75788-904
Human Recombinant Carbonic Anhydrase 1 (from E. coli)
Enzymes
Carbonic Anhydrase 1 (CA1) is a cytosolic enzyme, belonging to the alpha-carbonic anhydrase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is highly expressed in erythrocytes and acts as an early marker for erythroid differentiation. Carbonic anhydrase 1 plays a improtant role in many biological processes such as calcification, cellular respiration, bone resorption, acid-base balance. It is activated by imidazole, histamine, L-adrenaline, L- and D-histidine, and L- and D-phenylalanine. At the same time, It is inhibited by sulfonamide derivatives and coumarins. In addition, CA1 is a zinc metalloenzyme that has reversible hydration of carbon dioxide. It can hydrate cyanamide to urea.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty