Would you like to log in again?
Human Recombinant Carbonic Anhydrase 4 (from E. coli)
Supplier: ProSci
91-092
75788-908EA 946.47 USD InStock
75788-908
Human Recombinant Carbonic Anhydrase 4 (from E. coli)
Enzymes
Carbonic Anhydrase 4 (CA4) belongs to the alpha-carbonic anhydrase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Alpha-carbonic anhydrase is a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Carbonic anhydrase 4 is a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary (and certain other) capillaries and proximal renal tubules. Carbonic anhydrase 4 may stimulate the sodium/bicarbonate transporter activity of SLC4A4 that acts in pH homeostasis. It may have a role in inherited renal abnormalities of bicarbonate transport. Furthermore, Carbonic anhydrase 4 is essential for acid overload removal from the retina and retina epithelium and acid release in the choriocapillaris.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty