Would you like to log in again?
Human Recombinant Carbonic Anhydrase 8 (from E. coli)
Supplier: ProSci
91-095
75788-914EA 533.61 USD InStock
75788-914
Human Recombinant Carbonic Anhydrase 8 (from E. coli)
Enzymes
Carbonic Anhydrase 8 (CA8) belongs to the alpha-carbonic anhydrase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Alpha-carbonic anhydrase is a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Because CA8 has some sequence similarity with other known carbonic anhydrase genes, it was firstly called as CA-related protein. Nevertheless, CA8 does not have a carbonic anhydrase catalytic activity. Defects in CA8 are the cause of cerebellar ataxia mental retardation and dysequilibrium syndrome type 3 (CMARQ3), which is a congenital cerebellar ataxia associated with dysarthia, quadrupedal gait and mild mental retardation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty