Would you like to log in again?
Human Recombinant Esterase D (from E. coli)
Supplier: ProSci
91-115
75788-946EA 700.27 USD InStock
75788-946
Human Recombinant Esterase D (from E. coli)
Enzymes
Human Esterase D is a serine hydrolase that is involved in the detoxification of formaldehyde
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Esterase D plays a part in a variety of substrates, including O-acetylated sialic acids, which may involves in the recycling of sialic acids. Esterase D can be used as a genetic marker for retinoblastoma and Wilson’s disease.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty