Would you like to log in again?
Human Recombinant Fructose-1,6-Bisphosphatase 1 (from E. coli)
Supplier: ProSci
91-122
75788-960EA 644.75 USD InStock
75788-960
Human Recombinant Fructose-1,6-Bisphosphatase 1 (from E. coli)
Enzymes
Fructose-1,6-Bisphosphatase 1 (FBPase 1) is a member of the FBPase class 1 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
FBPase 1 is a gluconeogenesis regulatory protein, which catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. FBPase 1 can assume an active R-state, or an inactive T-state. FBPase 1 deficiency is inherited as an autosomal recessive disorder mainly in the liver and causes life-threatening episodes of hypoglycemia and metabolic acidosis in newborn infants or young children. FBPase 1 coupled with phosphofructokinase (PFK) is involved in the metabolism of pancreatic islet cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty