Would you like to log in again?
Human Recombinant DNA Polymerase beta (from E. coli)
Supplier: ProSci
91-139
75788-994EA 700.07 USD InStock
75788-994
Human Recombinant DNA Polymerase beta (from E. coli)
Enzymes
Human DNA polymerase beta is constitutively expressed in cells
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It fills in gaps in DNA that are formed following base excision repair. The activity cannot be affected by Aphidicolin, which is an inhibitor of DNA polymerase beta .
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty